Learn More
Jorgensen Labs Blunt Tip Needles, 22 g x 1", Blue, 5/pk
SDP
Catalog No. NC0502914
Encompass_Preferred
These blunt end needles have a wide range of applications around the clinic. These blunt ends have a polished straight stainless cannula with a polypropylene color-coded hub. Non-sterile 5 pack.
Catalog No. NC0502914 Supplier Jorgensen Labs Supplier No. J1032G
These blunt end needles have a wide range of applications around the clinic. These blunt ends have a polished straight stainless cannula with a polypropylene color-coded hub. Non-sterile 5 pack.