Learn More
Access Technologies Huber Point Non-Coring Needles, 22ga, 3/4” (1.6cm), Black
Catalog No. NC9721079
Encompass
Feature an architecturally contoured “B” bevel needle tip and a unique knarled hub. This hub provides positive grip action when attaching the needle to a syringe or luer lock. The PosiGrip hub is color coded for easy gauge identification.
Catalog No. NC9721079 Supplier Access Technologies Supplier No. PG2275
Feature an architecturally contoured “B” bevel needle tip and a unique knarled hub. This hub provides positive grip action when attaching the needle to a syringe or luer lock. The PosiGrip hub is color coded for easy gauge identification.