Learn More
Havels 22GX3.5 SPROTTE NDL W/INTRODU
SDP
Catalog No. NC0107679
Encompass
The Atraumatic Sprotte Spinal Needle for spinal anesthesia and diagnostic lumbar puncture reduces post-spinal headaches by a factor of ten! The conical point reduces dura trauma because it spreads dura fibers - it doesn't cut.
Catalog No. NC0107679 Supplier Havels Supplier No. INS2290
The Atraumatic Sprotte Spinal Needle for spinal anesthesia and diagnostic lumbar puncture reduces post-spinal headaches by a factor of ten! The conical point reduces dura trauma because it spreads dura fibers - it doesn't cut.