Learn More
MSC Ansell Glove, Size Regular, Yellow, ANSI Cut A2, 1 ea
Catalog No. 502886339
Encompass
Ansell abrasion- and cut-resistant gloves are designed to protect hands in various industrial applications. This glove offers ANSI Cut A2 protection, is yellow, and comes in a regular size.
  • Launderable
  • Covers from forearm to wrist
  • Lightweight
  • Comfortable
Catalog No. 50-288-6339 Supplier MSC Supplier No. 43699842
Ansell abrasion- and cut-resistant gloves are designed to protect hands in various industrial applications. This glove offers ANSI Cut A2 protection, is yellow, and comes in a regular size.
  • Launderable
  • Covers from forearm to wrist
  • Lightweight
  • Comfortable