Learn More
Thomas Scientific Drawer Rack combo 8 box, 39 50ml tubes
SDP
Catalog No. 50196040
Encompass
Rack w/2 Drawers for 4 Standard 2" Cardboard Boxes&I Drawer for Storage Bottles 8 Boxes Plus 80 Tubes (15ml) 8 Boxes Plus 39 Tubes (50ml) Outside Dimensions:22 x 9 11/16 x 5 1/2 inch in ( 559 x 246 x 140
Catalog No. 50-196-040 Supplier Thomas Scientific Supplier No. 1149N94
Rack w/2 Drawers for 4 Standard 2" Cardboard Boxes&I Drawer for Storage Bottles 8 Boxes Plus 80 Tubes (15ml) 8 Boxes Plus 39 Tubes (50ml) Outside Dimensions:22 x 9 11/16 x 5 1/2 inch in ( 559 x 246 x 140