Learn More
Thomas Scientific 22" Length- 2 Drawer Upright Freezer Rack for Bottles, 22" L x 9" h x 5 1/2" D, Max Bottle Height: 4 3/16"
SDP
Catalog No. 50196009
Encompass_Preferred
Thomas No. 1200C38, 1200C39 and 1200C40 Rack with 1 Drawer for Bottles Thomas No. 1200C41, 1200C42 and 1200C43 Rack with 2 Drawer for Bottles
Catalog No. 50-196-009 Supplier Thomas Scientific Supplier No. 1148X32
Thomas No. 1200C38, 1200C39 and 1200C40 Rack with 1 Drawer for Bottles Thomas No. 1200C41, 1200C42 and 1200C43 Rack with 2 Drawer for Bottles