Learn More
Simco 22IN PLSD/STDY STATE STNDLN
Catalog No. NC3353909
Encompass_Preferred
22" Pulsed & Steady State Standalone AeroBar (24VAC) with FMS Alarm Output and Titanium Emitter Points (5), RoHS. Requires Handheld Remote Below (91-5570) (No Interface Module Required)
Catalog No. NC3353909 Supplier Simco Supplier No. 915225SC22R
22" Pulsed & Steady State Standalone AeroBar (24VAC) with FMS Alarm Output and Titanium Emitter Points (5), RoHS. Requires Handheld Remote Below (91-5570) (No Interface Module Required)