Learn More
Grainger 22INX17X6IN DIVIDER BLUE
Catalog No. NC0797280
Encompass_Preferred
Divider Box, 22-3/8 x 17-3/8 x 6 In, Blue. Featuring industrial-grade polymer construction, this dividable grid container has slots to create multi-sized compartments. The molded-in letter/number system allows easy parts tracking. The reinforcing ribs and strong stacking rims provide strength, rigidity, and shape retention. Optional black intersecting dividers, available separately, allow subdivision to 1-1/8" square.
Catalog No. NC0797280 Supplier Grainger Supplier No. 2RV47
Divider Box, 22-3/8 x 17-3/8 x 6 In, Blue. Featuring industrial-grade polymer construction, this dividable grid container has slots to create multi-sized compartments. The molded-in letter/number system allows easy parts tracking. The reinforcing ribs and strong stacking rims provide strength, rigidity, and shape retention. Optional black intersecting dividers, available separately, allow subdivision to 1-1/8" square.