Learn More
Ace Glass, Inc. 22L Glas-Col mantle, aluminum housed, hemispherical, 3" bottom hole, (2) 770w - 115v heating
SDP
Catalog No. 50977306
Encompass
22L Glas-Col aluminum housed, hemispherical heating mantle with 3 holes in bottom to accomm ODate flasks with bottom outlets. 770w-115v heating element is embedded in layers of glass fabric to protect the flask wall from thermal strain, and is covered with glass insulation to prevent heat from radiating outward. Glass fabric affords temperatures to 450 C. Includes 4ft, 3-wire cord and locking connector.
Catalog No. 50-977-306 Supplier Ace Glass, Inc. Supplier No. 1204430
22L Glas-Col aluminum housed, hemispherical heating mantle with 3 holes in bottom to accomm ODate flasks with bottom outlets. 770w-115v heating element is embedded in layers of glass fabric to protect the flask wall from thermal strain, and is covered with glass insulation to prevent heat from radiating outward. Glass fabric affords temperatures to 450 C. Includes 4ft, 3-wire cord and locking connector.