Learn More
Ace Glass, Inc. 22mm quartz graded seals, 7.6 cm length
SDP
Catalog No. 50974601
Encompass
22mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-601 Supplier Ace Glass, Inc. Supplier No. 845534
22mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.