Learn More
Molecular Dimensions 22MM SQUARE COVER SLIPS (1000)
SDP
Catalog No. 501038557
Encompass
22mm square cover slips (1000) Non-Siiconized. No.2 size = 0.19 mm - 0.23 mm thickness- standard for crystallization.
Catalog No. 50-103-8557 Supplier Molecular Dimensions Supplier No. MD409
22mm square cover slips (1000) Non-Siiconized. No.2 size = 0.19 mm - 0.23 mm thickness- standard for crystallization.