Learn More
Ace Glass, Inc. 22mm x 30cm chromatography column
SDP
Catalog No. 50972548
Encompass_Preferred
22mm x 30cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-972-548 Supplier Ace Glass, Inc. Supplier No. 588837
22mm x 30cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.