Learn More
MSC Baldwin Filter, 12 1/8 x 23 1/2 in., 1 ea
Catalog No. 5000505016
Encompass
This automotive engine outer air filter is designed with a round shape, providing efficient filtration for automotive applications. It is an element-only filter, ensuring precise fit and performance.
  • Engine outer air filter type
  • Round design
  • Element only
  • Includes two attached gaskets
  • Measures 23 1/2 in. in height
  • Features 12 1/8 in. outside diameter
Catalog No. 50-005-05016 Supplier MSC Supplier No. 10939825
This automotive engine outer air filter is designed with a round shape, providing efficient filtration for automotive applications. It is an element-only filter, ensuring precise fit and performance.
  • Engine outer air filter type
  • Round design
  • Element only
  • Includes two attached gaskets
  • Measures 23 1/2 in. in height
  • Features 12 1/8 in. outside diameter