Learn More
Arbor Assays 2,3-cGAMP STING-based FRET Detection Kits (5 Plate)
SDP
Supplier:  Arbor Assays K081F5
Encompass_Preferred
The DetectX 2,3-cGAMP STING-Based FRET Kits quantitatively measure 2,3-cGAMP in lysed cells and tissue, as well as tissue culture media samples.
Catalog No. 50-227-8526