Learn More
Justrite™ 23 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822343 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-343 Light Neutral
19-822-345 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-343 Supplier Justrite™ Supplier No. 883627
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 23 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Keep personnel and parts away from fluids and contaminants
  • Durable, anti-slip grates help keep personnel and other objects above contaminated wash down water
  • Made of chemical resistant, easy-to-clean polyethylene
  • Service temperature range: -30F to 120F
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Door Style Self-close
Color Light Neutral
Load Bearing Capacity 23 gal.