Learn More
Braintree Scientific Blunt Tip Needle Non-Sterile, 23ga - 1" w/std hub, 12/pk
SDP
Catalog No. NC9250449
Encompass
Blunt Tip Needle, 23 Gauge 1 in. Stainless Steel Hypodermic Needle with Standard Plastic Luer Hub, used to Connect Syringe Directly to Catheter Port for Intravascular Substance Administration, Blood Sampling and Catheter Flushing **Sold for research purposes only and not for human use**
Catalog No. NC9250449 Supplier Braintree Scientific Supplier No. NB23G
Blunt Tip Needle, 23 Gauge 1 in. Stainless Steel Hypodermic Needle with Standard Plastic Luer Hub, used to Connect Syringe Directly to Catheter Port for Intravascular Substance Administration, Blood Sampling and Catheter Flushing **Sold for research purposes only and not for human use**