Learn More
Cayman Chemical 23-hydroxy Betulinic Acid, Purity: ≥98%, CAS: 85999-40-2, 5 mg
SDP
Catalog No. 502938720
Encompass_Preferred
A triterpenoid with anticancer activity; inhibits proliferation of K562, B16, and HeLa cells (IC50s = 39.9, 78.5, and 80 µM, respectively), as well as HUVECs (IC50 = 94.8 µM); halts the cell cycle at the S phase and induces apoptosis in K562 cells
Catalog No. 50-293-8720 Supplier Cayman Chemical Supplier No. 274465 MG
A triterpenoid with anticancer activity; inhibits proliferation of K562, B16, and HeLa cells (IC50s = 39.9, 78.5, and 80 µM, respectively), as well as HUVECs (IC50 = 94.8 µM); halts the cell cycle at the S phase and induces apoptosis in K562 cells