Learn More
LUKNOVA INC C18, Octadecyl-Functionalized Silica Gel, Reversed Phase Material. Organic Loading 17%, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å And Endcapped. 1 Kg Per Pack. Standard C18-Bonded On High Purity Silica Gel For Chromatography Application.
Catalog No. 50920656
Encompass
C18, Octadecyl-Functionalized Silica Gel, Reversed Phase Material. Organic Loading 17%, Particle Size 40-63 µm (230-400 Mesh), And Pore Size 60 Å. Endcapped To Enable Additional Non-Polar Retention By Minimizing Polar Silanol Interactions. This Unit Comes At 1 Kg Package Of Plastic Bottle. Standard C18-Bonded On High Purity Irregular Shape Silica Gel With Narrow Particle Size Distribution To Allow Great Chromatography Purification Efficiency, Resolution, Reproducibility, Loading Capacity And Recovery Rates. Used For Reversed Phase Chromatography For Extraction Of Most Non-Polar Organic Analytes From Aqueous And Biological Samples, Like Glycerides, Fatty Acids, Polycyclic Aromatics, Esters (Phthalates), Fat-Soluble Vitamins, Steroids, Prostaglandins, Pth Amino Acids, Et Al.
Catalog No. 50-920-656 Supplier LUKNOVA INC Supplier No. SGFLASHC181
C18, Octadecyl-Functionalized Silica Gel, Reversed Phase Material. Organic Loading 17%, Particle Size 40-63 µm (230-400 Mesh), And Pore Size 60 Å. Endcapped To Enable Additional Non-Polar Retention By Minimizing Polar Silanol Interactions. This Unit Comes At 1 Kg Package Of Plastic Bottle. Standard C18-Bonded On High Purity Irregular Shape Silica Gel With Narrow Particle Size Distribution To Allow Great Chromatography Purification Efficiency, Resolution, Reproducibility, Loading Capacity And Recovery Rates. Used For Reversed Phase Chromatography For Extraction Of Most Non-Polar Organic Analytes From Aqueous And Biological Samples, Like Glycerides, Fatty Acids, Polycyclic Aromatics, Esters (Phthalates), Fat-Soluble Vitamins, Steroids, Prostaglandins, Pth Amino Acids, Et Al.