Learn More
LUKNOVA INC High Purity Silica Gel, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å, Ph Range 6.8-7.2. This Unit Comes At 25 Kg Per Package In Plastic Drum. Standard Silica Gel Has Irregular Shape And Great For Chromatography Application.
Catalog No. 50920653
Encompass
High Purity Silica Gel, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å, Pore Volume 0.75 Ml/G, Surface Area 500M2/G, Ph Range 6.8-7.2. This Unit Comes At 25 Kg Per Package In Plastic Drum. This Standard Silica Gel Has Irregular Shape And Free-Flowing Fine Powder Form And Great For Flash Chromatography Application With Sample Loading Up To 10-Wt%. It Has Narrow Particle Size Distribution And Narrow Ph Range Which Enable Significant Advantages In Purification Efficiency, Resolution, And Column Reproducibility. Cas Number 112926-00-8.
Catalog No. 50-920-653 Supplier LUKNOVA INC Supplier No. SG23040025
High Purity Silica Gel, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å, Pore Volume 0.75 Ml/G, Surface Area 500M2/G, Ph Range 6.8-7.2. This Unit Comes At 25 Kg Per Package In Plastic Drum. This Standard Silica Gel Has Irregular Shape And Free-Flowing Fine Powder Form And Great For Flash Chromatography Application With Sample Loading Up To 10-Wt%. It Has Narrow Particle Size Distribution And Narrow Ph Range Which Enable Significant Advantages In Purification Efficiency, Resolution, And Column Reproducibility. Cas Number 112926-00-8.