Learn More
LUKNOVA INC High Purity Silica Gel, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å, Ph Range 6.8-7.2. This Unit Comes At 500 Gram Per Bottle. Standard Silica Gel Has Irregular Shape And Great For Chromatography Application.
Catalog No. 50920669
Encompass
High Purity Silica Gel, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å, Pore Volume 0.75 Ml/G, Surface Area 500M2/G, Ph Range 6.8-7.2. This Unit Comes At 500 Gram Per Plastic Bottle. This Standard Silica Gel Has Irregular Shape And Free-Flowing Fine Powder Form And Great For Flash Chromatography Application With Sample Loading Up To 10-Wt%. It Has Narrow Particle Size Distribution And Narrow Ph Range Which Enable Significant Advantages In Purification Efficiency, Resolution, And Column Reproducibility. Cas Number 112926-00-8.
Catalog No. 50-920-669 Supplier LUKNOVA INC Supplier No. SG23040005
High Purity Silica Gel, Particle Size 40-63 µm (230-400 Mesh), Pore Size 60 Å, Pore Volume 0.75 Ml/G, Surface Area 500M2/G, Ph Range 6.8-7.2. This Unit Comes At 500 Gram Per Plastic Bottle. This Standard Silica Gel Has Irregular Shape And Free-Flowing Fine Powder Form And Great For Flash Chromatography Application With Sample Loading Up To 10-Wt%. It Has Narrow Particle Size Distribution And Narrow Ph Range Which Enable Significant Advantages In Purification Efficiency, Resolution, And Column Reproducibility. Cas Number 112926-00-8.