Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BACTEC™ Plus Aerobic Medium

Catalog No. 23025906 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-025-906 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-025-906 Supplier BD Supplier No. 442023
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

Support the growth of common aerobic organisms

  • An all-purpose, enriched soybean-casein digest broth medium capable of supporting the growth of aerobic organisms
  • Contains resins for antibiotic neutralization
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
No. per Pack 50
Packaging Type Plastic Culture Vials
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.