Learn More
Labtech Inc Recirculating Water Chiller 1000w
Catalog No. 501959346
Encompass_Preferred
Recirculating Water Chiller by LabTech, cooling capacity of 1000 Watt at 25˚C, 230V, 8C ~ 35˚C, model H150-1000-230
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 50-195-9346 Supplier Labtech Inc Supplier No. LW1000230V
Recirculating Water Chiller by LabTech, cooling capacity of 1000 Watt at 25˚C, 230V, 8C ~ 35˚C, model H150-1000-230
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.