Learn More
MSC Tape, 3M Masking, 1 in, 60 yd, Tan, 1 Roll
Catalog No. 501150097
Encompass
This multipurpose masking tape is crepe-backed with a rubber adhesive, designed for indoor applications. It performs well in general painting and paint bake operations up to 250°F (121°C) for 30 minutes, and is suitable for holding, protecting, and bundling.
  • Strong backing resists slivering and provides one-piece removal
  • Rubber adhesive provides instant adhesion and resists lifting or curling
  • Crepe paper backing conforms and maintains integrity around corners
  • Manufactured without solvents for reduced environmental impact
  • Controlled unwind ensures even dispensing, reducing premature tearing
  • Bonds well to paper, metal, wood, and painted surfaces
Catalog No. 50-115-0097 Supplier MSC Supplier No. 77185098
This multipurpose masking tape is crepe-backed with a rubber adhesive, designed for indoor applications. It performs well in general painting and paint bake operations up to 250°F (121°C) for 30 minutes, and is suitable for holding, protecting, and bundling.
  • Strong backing resists slivering and provides one-piece removal
  • Rubber adhesive provides instant adhesion and resists lifting or curling
  • Crepe paper backing conforms and maintains integrity around corners
  • Manufactured without solvents for reduced environmental impact
  • Controlled unwind ensures even dispensing, reducing premature tearing
  • Bonds well to paper, metal, wood, and painted surfaces