Learn More
MSC 234 36MMX55M NATURAL MED-TEMP
Catalog No. 501297452
Encompass
The 3M General Purpose Masking Tape 234 is a crepe-backed masking tape designed for general painting and paint bake operations. It features a rubber adhesive and can perform reliably in temperatures up to 250°F (121°C).
  • Strong backing resists slivering and allows one-piece removal.
  • Rubber adhesive provides instant adhesion and resists lifting or curling.
  • Crepe paper backing conforms and maintains integrity around corners.
  • Performs well in paint bake operations up to 250°F (121°C) for 30 minutes.
  • Manufactured without solvents for reduced environmental impact.
  • Removes cleanly and easily from most surfaces.
  • Moderate conformability over irregular surfaces.
  • Bonds well to paper, metal, wood, and painted surfaces.
Catalog No. 50-129-7452 Supplier MSC Supplier No. 02999506
The 3M General Purpose Masking Tape 234 is a crepe-backed masking tape designed for general painting and paint bake operations. It features a rubber adhesive and can perform reliably in temperatures up to 250°F (121°C).
  • Strong backing resists slivering and allows one-piece removal.
  • Rubber adhesive provides instant adhesion and resists lifting or curling.
  • Crepe paper backing conforms and maintains integrity around corners.
  • Performs well in paint bake operations up to 250°F (121°C) for 30 minutes.
  • Manufactured without solvents for reduced environmental impact.
  • Removes cleanly and easily from most surfaces.
  • Moderate conformability over irregular surfaces.
  • Bonds well to paper, metal, wood, and painted surfaces.