Learn More
Uline 3M 234 High Temperature Masking Tape - 2"" x 60 yds | 12/pack
Catalog No. NC1892173
Encompass
Tough industrial tape for paint masking and bundling. Adheres to stainless steel, powder coatings, EPDM rubber and alodine aluminum. Good paint line. Clean removal. Resists solvents and moisture. Withstands 250¿F for up to 30 minutes. Smooth tan crepe paper backing with rubber adhesive.
Catalog No. NC1892173 Supplier Uline Supplier No. S5699PK
Tough industrial tape for paint masking and bundling. Adheres to stainless steel, powder coatings, EPDM rubber and alodine aluminum. Good paint line. Clean removal. Resists solvents and moisture. Withstands 250¿F for up to 30 minutes. Smooth tan crepe paper backing with rubber adhesive.