Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 2360 Series P100 Particulate Respirator with Ventex™ Valve
SDP

Catalog No. 19059332 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-059-332 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-059-332 Supplier Moldex™ Supplier No. 2360P100

P100 particulate respirator with Ventex exhalation valve allows hot air to escape faster to give wearer a cooler and easier breathing experience.

Application

  • Use in environments where an OSHA substance-specific standard applies (lead, cadmium, arsenic, MDA) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
  • Design
    • Adjustable cloth straps assure snug, comfortable fit
    • Ventex valve lets the hot air out faster so workers breathe easier and stay cooler
    • Soft foam face cushion for added comfort and no pressure points
    • Dura-Mesh™ shell resists collapsing in regular use and even under hot and humid conditions
    • Contour-molded nose bridge seals without metal noseband
    • Single use

    Do not use against concentrations of particulates that exceed the maximum use concentration or 10X the OSHA PEL, whichever is lower.

    Specifications

    Aerosol Type P - Oil-Proof
    ANSI Standard ANSI/ISEA 110-2003, Section 7.11.1
    Certifications (NIOSH) NIOSH 42 CFR 84, P100
    Color White
    Design Type Cup/Molded
    Disposable Single-use
    Filter Type P100
    Material Dura-Mesh
    Product Type General Purpose Particulate Respirator
    Size Medium/Large
    With Exhalation Valve Yes
    Filter Efficiency 99%
    For Use With (Application) Environments where an OSHA substance specific standard applies (lead, cadmium, arsenic, mda) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
    Model 2360P100
    Nosepiece Type Molded Nose Bridge
    Quantity 5/Pk.
    Strap Attachment Type Adjustable Cloth Straps
    FDA Cleared Surgical Mask No
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.