Learn More
Medchemexpress LLC PBT434 mesylate | 2387898-69-1 | 25 MG
SDP
Catalog No. 5000234172
Encompass_Preferred
PBT434 mesylate is the methanesulfonate salt of PBT434, a brain-penetrant, orally active small molecule inhibitor of α-synuclein aggregation used in preclinical Parkinson's disease research.
  • Acts as an iron chelator and modulates transcellular iron trafficking.
  • Inhibits iron-mediated redox activity and α-synuclein aggregation.
  • Orally active and crosses the blood-brain barrier.
  • Supplied as a mesylate salt, solid, white to off-white.
  • High purity suitable for research (99.9%).
Catalog No. 50-002-34172 Supplier Medchemexpress LLC Supplier No. HY120475A25MG
PBT434 mesylate is the methanesulfonate salt of PBT434, a brain-penetrant, orally active small molecule inhibitor of α-synuclein aggregation used in preclinical Parkinson's disease research.
  • Acts as an iron chelator and modulates transcellular iron trafficking.
  • Inhibits iron-mediated redox activity and α-synuclein aggregation.
  • Orally active and crosses the blood-brain barrier.
  • Supplied as a mesylate salt, solid, white to off-white.
  • High purity suitable for research (99.9%).