Learn More
SAI Infusion Technologies 23 Gauge, 1.5" Blunt Needles - Sterile, 25/pk
SDP
Catalog No. NC9031029
Encompass
23G1.5" Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. Blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters. SAI's smooth, burr-free stainless steel needles prevent damage to the lumen of the catheter during dosing, sampling, flushing or locking. These needles are sterile and individually packaged, 25/pk. Non-sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC9031029 Supplier SAI Infusion Technologies Supplier No. B23150
23G1.5" Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. Blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters. SAI's smooth, burr-free stainless steel needles prevent damage to the lumen of the catheter during dosing, sampling, flushing or locking. These needles are sterile and individually packaged, 25/pk. Non-sterile blunt needles are available separately. Product is for laboratory animal research only.