Learn More
SAI Infusion Technologies Non-Sterile Blunt Needles, 23G, 0.5"", 100/pack
Supplier: SAI Infusion Technologies B2350 100B
This item has a minimum qty of 2 per supplier requirements.
23G0.5 Blunt Needles with plastic hub and stainless steel blunt needle. Blunt tip needles are useful for a variety of laboratory tasks, including connection to indwelling catheters. SAI's smooth, burr-free stainless steel needles prevent damage to the lumen of the catheter during dosing, sampling, flushing or locking. These needles are non-sterile, 100/pk. ETO Exposed blunt needles also available. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **