Learn More
SAI Infusion Technologies Stainless Steel Catheter Plugs - 23G, 1cm, 100/pk
SDP
Catalog No. NC9995642
Encompass_Preferred
SAI's Stainless Steel Catheter Plugs safely seal catheters when not in use. Burr-free stainless steel prevents damage to the catheter during insertion and removal. Catheter pins are solid. To connect two catheters instead, use hollow steel catheter connectors (CC-23). Non-sterile, 23G, 1cm, 100/pk. Product is for laboratory animal research only.
Catalog No. NC9995642 Supplier SAI Infusion Technologies Supplier No. CP23
SAI's Stainless Steel Catheter Plugs safely seal catheters when not in use. Burr-free stainless steel prevents damage to the catheter during insertion and removal. Catheter pins are solid. To connect two catheters instead, use hollow steel catheter connectors (CC-23). Non-sterile, 23G, 1cm, 100/pk. Product is for laboratory animal research only.