Learn More
SAI Infusion Technologies Stainless Steel Catheter Connectors - 23G, 1cm
SDP
Catalog No. NC9825557
Encompass
SAI's catheter connectors ensure a safe and effective fit when connecting fluid lines for seamless fluid flow. Burr-free stainless steel prevents damage to the catheter during insertion and removal. 23G, 1cm, 100/pk. To plug a catheter, use solid stainless steel catheter pins (CP-23). Product is for laboratory animal research only.
Catalog No. NC9825557 Supplier SAI Infusion Technologies Supplier No. CC23
SAI's catheter connectors ensure a safe and effective fit when connecting fluid lines for seamless fluid flow. Burr-free stainless steel prevents damage to the catheter during insertion and removal. 23G, 1cm, 100/pk. To plug a catheter, use solid stainless steel catheter pins (CP-23). Product is for laboratory animal research only.