Learn More
Medchemexpress LLC 24:0 L-carnitine-d4 | 2260670-68-4 | 515.84 | 1 MG

Supplier: Medchemexpress LLC HY146943S1MG
24:0 L-carnitine-d4 is a chemical compound identified by CAS Number 2260670-68-4 and a molecular weight of 515.84. This product is generally used in research and scientific applications.
- Recommended for long-term storage at -80°C
- Recommended for shorter storage periods at -20°C
- Store in sealed containers
- Protect from moisture and light
- Avoid repeated freeze-thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.