Learn More
Medchemexpress LLC 24:0 Lyso PC-13C6 | 2483735-01-7 | 97.5% | 613.80 | 1 MG
SDP
Catalog No. 5000351680
Encompass_Preferred
24:0 Lyso PC-13C6 is the 13C labeled 24:0 Lyso PC. 24:0 Lyso PC is a lysophospholipid (LyP) that could be used for mRNA drug delivery. It can also be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-51680 Supplier Medchemexpress LLC Supplier No. HY138622S1MG
24:0 Lyso PC-13C6 is the 13C labeled 24:0 Lyso PC. 24:0 Lyso PC is a lysophospholipid (LyP) that could be used for mRNA drug delivery. It can also be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS