Learn More
Cayman Chemical 24 25-epoxy Cholesterol 1mg
SDP
Catalog No. 502937017
Encompass_Preferred
An oxysterol; inhibits the proliferation of HGC-27 cells at 1 µM, an effect that can be enhanced by knockdown of LXRβ; tumor levels are increased in patients with gastric cancer
Catalog No. 50-293-7017 Supplier Cayman Chemical Supplier No. 259661 MG
An oxysterol; inhibits the proliferation of HGC-27 cells at 1 µM, an effect that can be enhanced by knockdown of LXRβ; tumor levels are increased in patients with gastric cancer