Learn More
Ace Glass, Inc. 24/25 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Catalog No. 50977608
Encompass
24/25 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-608 Supplier Ace Glass, Inc. Supplier No. 1263616
24/25 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.