Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 24/40 outer top, 10/30inner bottom
SDP
Catalog No. 50971592
Encompass_Preferred
Connecting, enlarging adapter featuring a 24/40 outer top female joint and a 10/30 inner bottom male joint.
This item is not returnable. View return policy
Catalog No. 50-971-592 Supplier Ace Glass, Inc. Supplier No. 500508
Connecting, enlarging adapter featuring a 24/40 outer top female joint and a 10/30 inner bottom male joint.