Learn More
Ace Glass, Inc. 10.5mm ID x 25cm chromatography column, 24/40 outer top, 200mL reservoir, without filter disc
SDP
Supplier:  Ace Glass, Inc. 5907105
Encompass_Preferred
10.5mm I.D. x 25cm chromatography column with 200mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint.
Catalog No. 50-972-594