Learn More
Ace Glass, Inc. 24/40 10mL Dean & Stark Moisture Test Receiver
SDP
Catalog No. 50975336
Encompass_Preferred
24/40 10mL Dean & Stark Moisture Test Receiver with return line to allow excess solvent to return to the boiling flask. Suitable for ASTM D 95 and E 123.
Catalog No. 50-975-336 Supplier Ace Glass, Inc. Supplier No. 772502
24/40 10mL Dean & Stark Moisture Test Receiver with return line to allow excess solvent to return to the boiling flask. Suitable for ASTM D 95 and E 123.