Learn More
Ace Glass, Inc. 24/40 10mm debris trap, glass only
SDP
Catalog No. 50979645
Encompass_Preferred
24/40 10mm glass portion for PTFE Trubore debris trap. Use with ACE pstainless steel or polished glass 10mm O.D. stir shafts.
Catalog No. 50-979-645 Supplier Ace Glass, Inc. Supplier No. 8066220
24/40 10mm glass portion for PTFE Trubore debris trap. Use with ACE pstainless steel or polished glass 10mm O.D. stir shafts.