Learn More
Ace Glass, Inc. Complete 24/40 10mm bearing, straight adapter style with hose connection port
SDP
Catalog No. 50975968
Encompass_Preferred
24/40 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-975-968 Supplier Ace Glass, Inc. Supplier No. 8066130
24/40 10mm self-aligning, lubricant-free PTFE Trubore straight adapter style bearing with hose connection port. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.