Learn More
Ace Glass, Inc. Connecting adapter, reducing, 24/40 outer top, 12/30 inner bottom
SDP
Catalog No. 50971593
Encompass_Preferred
Connecting adapter featuring a 24/40 outer top female joint and a 12/30 inner bottom male joint.
Catalog No. 50-971-593 Supplier Ace Glass, Inc. Supplier No. 500510
Connecting adapter featuring a 24/40 outer top female joint and a 12/30 inner bottom male joint.