Learn More
Ace Glass, Inc. Connecting adapter with 24/40 top female and 14/20 bottom male standard taper joints.
SDP
Catalog No. 509558655
Encompass_Preferred
Connecting adapter with 24/40 top female and 14/20 bottom male standard taper joints.
Catalog No. 50-955-8655 Supplier Ace Glass, Inc. Supplier No. 909226
Connecting adapter with 24/40 top female and 14/20 bottom male standard taper joints.