Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 24/40 outer top, 14/35 inner bottom
SDP
Catalog No. 50971594
Encompass_Preferred
Connecting, enlarging adapter featuring a 24/40 outer top female joint and a 14/35 inner bottom male joint.
Catalog No. 50-971-594 Supplier Ace Glass, Inc. Supplier No. 500512
Connecting, enlarging adapter featuring a 24/40 outer top female joint and a 14/35 inner bottom male joint.