Learn More
Ace Glass, Inc. 14.5mm ID x 25cm chromatography column, 24/40 outer top, 250mL reservoir, without filter disc
SDP
Catalog No. 50972595
Encompass_Preferred
14.5mm I.D. x 25cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint.
Catalog No. 50-972-595 Supplier Ace Glass, Inc. Supplier No. 5907110
14.5mm I.D. x 25cm chromatography column with 250mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a 24/40 outer joint.