Learn More
Chemglass Life Sciences Adapter, Bushing, PTFE, 24/40 Lower Inner, 19/38 Top Outer
SDP
Catalog No. CG1016T02
Encompass_Preferred
Adapter, Bushing, PTFE, 24/40 Lower Inner, 19/38 Top Outer. Similar to CG1016 series of adapters, but made entirely of PTFE.
Catalog No. CG1016T02 Supplier Chemglass Life Sciences Supplier No. CG1016T02
Adapter, Bushing, PTFE, 24/40 Lower Inner, 19/38 Top Outer. Similar to CG1016 series of adapters, but made entirely of PTFE.