Learn More
Ace Glass, Inc. 24/40 20.3cm distillation column
SDP
Catalog No. 50973273
Encompass_Preferred
Distillation column with 24/40 standard taper joints and perforated glass disc for packing support. 20.3cm length refers to the length from the packing support to the bottom of the outer ground joint.
Catalog No. 50-973-273 Supplier Ace Glass, Inc. Supplier No. 657304
Distillation column with 24/40 standard taper joints and perforated glass disc for packing support. 20.3cm length refers to the length from the packing support to the bottom of the outer ground joint.