Learn More
Ace Glass, Inc. 24/40 20ML MOISTURE RCVR 24/40 20ML MOISTURE RCVR
SDP
Catalog No. 50979437
Encompass_Preferred
20mL moisture receiver with 24/40 joint. Includes a 120¿, 1:5 PTFE stopcock which provides adequate shut-off in the intermediate position. Thermometer joint is 10/30.
Catalog No. 50-979-437 Supplier Ace Glass, Inc. Supplier No. 774702
20mL moisture receiver with 24/40 joint. Includes a 120¿, 1:5 PTFE stopcock which provides adequate shut-off in the intermediate position. Thermometer joint is 10/30.