Learn More
Ace Glass, Inc. 24/40 250mL distillation receiver
SDP
Catalog No. 50978842
Encompass
250mL borosilicate glass jacketed distilling receiver with 24/40 joint has vacuum manifold for removal of the sample during operation, without disturbing the system. Manifold stopcocks are 4mm bore, and receiver stopcock is 3mm bore. Condenser hose connections are size E, for use with 9.5mm (3/8) or 11.1mm (7/16) I.D. tubing. Manifold has 10mm O.D. olive-style connections.
Catalog No. 50-978-842 Supplier Ace Glass, Inc. Supplier No. 664208
250mL borosilicate glass jacketed distilling receiver with 24/40 joint has vacuum manifold for removal of the sample during operation, without disturbing the system. Manifold stopcocks are 4mm bore, and receiver stopcock is 3mm bore. Condenser hose connections are size E, for use with 9.5mm (3/8) or 11.1mm (7/16) I.D. tubing. Manifold has 10mm O.D. olive-style connections.