Learn More
Ace Glass, Inc. 24/40 250mL distillation receiver
SDP
Supplier:  Ace Glass, Inc. 663708
Encompass
250mL all-purpose receiver for general distillation work. Equipped with a complete stopcock manifold to allow contents of the receiver to be removed to a single receiving flask without disturbing the vacuum of the system
Catalog No. 50-978-841