Learn More
Ace Glass, Inc. 24/40 25mL Dean & Stark Moisture Test Receiver
SDP
Catalog No. 50975337
Encompass_Preferred
24/40 25mL Dean & Stark Moisture Test Receiver with return line to allow excess solvent to return to the boiling flask. Suitable for ASTM D 95 and E 123.
Catalog No. 50-975-337 Supplier Ace Glass, Inc. Supplier No. 772504
24/40 25mL Dean & Stark Moisture Test Receiver with return line to allow excess solvent to return to the boiling flask. Suitable for ASTM D 95 and E 123.